Tisk…

Thiazolo[4,5-c]pyridine 95%

Dodavatel: Apollo Scientific

Synonyms: thiazolo[4,5-c]pyridine

OR903144-1G OR903144-250MG
APOSOR903144-1GEA 3110 CZK
APOSOR903144-1G APOSOR903144-250MG
Thiazolo[4,5-c]pyridine 95%
Thiazolo[4,5-c]pyridine
Vzorec: C₆H₄N₂S
MW: 136,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13179628
CAS číslo: 273-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR