Methyl piperidine-1-carboxylate 98%
Dodavatel: Apollo Scientific
OR903069-25G
OR903069-5G
APOSOR903069-25GEA
8500
CZK
APOSOR903069-25G
APOSOR903069-5G
Methyl piperidine-1-carboxylate 98%
Methyl piperidine-1-carboxylate
Vzorec:
C₇H₁₃NO₂ MW: 143,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09991718 CAS číslo: 1796-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...