1-(tert-Butoxy)-2-nitrobenzene 98%
Dodavatel: Apollo Scientific
OR902860-1G
OR902860-5G
APOSOR902860-1GEA
6220
CZK
APOSOR902860-1G
APOSOR902860-5G
1-(tert-Butoxy)-2-nitrobenzene 98%
1-(tert-Butoxy)-2-nitrobenzene
Vzorec:
C₁₀H₁₃NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09909350 CAS číslo: 83747-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...