Tisk…

2-(2-Methoxyphenyl)piperidine 95%

Dodavatel: Apollo Scientific

OR902855-100MG OR902855-1G OR902855-250MG
APOSOR902855-100MGEA 5200 CZK
APOSOR902855-100MG APOSOR902855-1G APOSOR902855-250MG
2-(2-Methoxyphenyl)piperidine 95%
2-(2-Methoxyphenyl)piperidine
Vzorec: C₁₂H₁₇NO
MW: 191,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663553
CAS číslo: 118577-00-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR