Tisk…

2-Sec-butylpyrrolidine 95%

Dodavatel: Apollo Scientific

OR902826-100MG OR902826-1G OR902826-250MG
APOSOR902826-100MGEA 5390 CZK
APOSOR902826-100MG APOSOR902826-1G APOSOR902826-250MG
2-Sec-butylpyrrolidine 95%
2-Sec-butylpyrrolidine
Vzorec: C₈H₁₇N
MW: 127,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663436
CAS číslo: 383127-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR