2-Isobutylpiperidine hydrochloride 95%
Dodavatel: Apollo Scientific
OR902736-100MG
OR902736-1G
OR902736-250MG
APOSOR902736-100MGEA
5390
CZK
APOSOR902736-100MG
APOSOR902736-1G
APOSOR902736-250MG
2-Isobutylpiperidine hydrochloride 95%
2-Isobutylpiperidine hydrochloride
Vzorec:
C₉H₁₉N·ClH MW: 177,715 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07357608 CAS číslo: 14162-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...