(3-Piperidinophenyl)methanol 97%
Dodavatel: Apollo Scientific
OR902626-1G
OR902626-5G
APOSOR902626-1GEA
5200
CZK
APOSOR902626-1G
APOSOR902626-5G
(3-Piperidinophenyl)methanol 97%
(3-Piperidinophenyl)methanol
Vzorec:
C₁₂H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD07772816 CAS číslo: 852180-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...