Tisk…

5-Cyclopropyl-1H-pyrazole-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR902612-1G OR902612-5G
APOSOR902612-1GEA 5200 CZK
APOSOR902612-1G APOSOR902612-5G
5-Cyclopropyl-1H-pyrazole-4-carboxylic acid 95%
5-Cyclopropyl-1H-pyrazole-4-carboxylic acid
Vzorec: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06208645
CAS číslo: 870704-26-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR