Tisk…

Ethyl 6-(chloromethyl)pyridine-2-carboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 6-(chloromethyl)picolinate

OR902572-1G
APOSOR902572-1GEA 4530 CZK
APOSOR902572-1G
Ethyl 6-(chloromethyl)pyridine-2-carboxylate 97%
Ethyl 6-(chloromethyl)pyridine-2-carboxylate
Vzorec: C₉H₁₀ClNO₂
MW: 199,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878512
CAS číslo: 49668-99-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR