Tisk…

1-(2-Isocyanoethyl)-pyrrolidine 95%

Dodavatel: Apollo Scientific

OR902445-1G OR902445-250MG
APOSOR902445-1GEA 18420 CZK
APOSOR902445-1G APOSOR902445-250MG
1-(2-Isocyanoethyl)-pyrrolidine 95%
1-(2-Isocyanoethyl)-pyrrolidine
Vzorec: C₇H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD05222853
CAS číslo: 2920-10-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR