Tisk…
Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate 98%
Dodavatel: Apollo Scientific
OR902301-1G OR902301-250MG OR902301-5G
APOSOR902301-1GEA 4300 CZK
APOSOR902301-1G APOSOR902301-250MG APOSOR902301-5G
Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate 98%
Methyl 4-amino-1-methyl-1H-pyrazole-3-carboxylate
Vzorec: C₆H₉N₃O₂
MW: 155,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969986
CAS číslo: 637336-53-9