1-Methyl-4-(3-methylpyridin-2-yl)piperazine 96%
Dodavatel: Apollo Scientific
OR902129-1G
OR902129-5G
APOSOR902129-1GEA
4770
CZK
APOSOR902129-1G
APOSOR902129-5G
1-Methyl-4-(3-methylpyridin-2-yl)piperazine 96%
1-Methyl-4-(3-methylpyridin-2-yl)piperazine
Vzorec:
C₁₁H₁₇N₃ MW: 191,273 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12546501 CAS číslo: 1187386-43-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...