Tisk…

2-(Isopropylamino)nicotinonitrile 98%

Dodavatel: Apollo Scientific

OR902128-1G OR902128-25G OR902128-5G
APOSOR902128-1GEA 3540 CZK
APOSOR902128-1G APOSOR902128-25G APOSOR902128-5G
2-(Isopropylamino)nicotinonitrile 98%
2-(Isopropylamino)nicotinonitrile
Vzorec: C₉H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD09939041
CAS číslo: 28721-27-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR