Tisk…

Ethyl 4-hydroxynicotinate 98%

Dodavatel: Apollo Scientific

OR901985-1G OR901985-250MG
APOSOR901985-1GEA 14290 CZK
APOSOR901985-1G APOSOR901985-250MG
Ethyl 4-hydroxynicotinate 98%
Ethyl 4-hydroxynicotinate
Vzorec: C₈H₉NO₃
MW: 167.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234426
CAS číslo: 57905-31-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR