Tisk…

Ethyl 4-ethoxynicotinate 98%

Dodavatel: Apollo Scientific

OR901969-1G OR901969-5G
APOSOR901969-1GEA 6420 CZK
APOSOR901969-1G APOSOR901969-5G
Ethyl 4-ethoxynicotinate 98%
Ethyl 4-ethoxynicotinate
Vzorec: C₁₀H₁₃NO₃
MW: 195,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972185
CAS číslo: 33279-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR