Tisk…

(4-nitropyridin-2-yl)methanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(Hydroxymethyl)-4-nitropyridine

OR901944-1G OR901944-250MG
APOSOR901944-1GEA 3610 CZK
APOSOR901944-1G APOSOR901944-250MG
(4-nitropyridin-2-yl)methanol 95%
(4-nitropyridin-2-yl)methanol
Vzorec: C₆H₆N₂O₃
MW: 154,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774099
CAS číslo: 98197-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR