Tisk…

tert-Butyl 4-oxo-2,3-dihydroquinoline-1-carboxylate 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Boc-3,4-dihydroquinoline-4(2H)-one

OR901923-1G OR901923-5G
APOSOR901923-1GEA 2980 CZK
APOSOR901923-1G APOSOR901923-5G
tert-Butyl 4-oxo-2,3-dihydroquinoline-1-carboxylate 96%
tert-Butyl 4-oxo-2,3-dihydroquinoline-1-carboxylate
Vzorec: C₁₄H₁₇NO₃
MW: 247,29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697721
CAS číslo: 179898-00-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR