Tisk…

4-Methoxy-2-phenylpyridine 97%

Dodavatel: Apollo Scientific

OR901887-1G OR901887-5G
APOSOR901887-1GEA 6220 CZK
APOSOR901887-1G APOSOR901887-5G
4-Methoxy-2-phenylpyridine 97%
4-Methoxy-2-phenylpyridine
Vzorec: C₁₂H₁₁NO
MW: 185.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702090
CAS číslo: 53698-56-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR