Tisk…

3-Acetamido-6-chloropyridazine 98%

Dodavatel: Apollo Scientific

Synonyms: N-(6-Chloropyridazin-3-yl)acetamide

OR901881-5G
APOSOR901881-5GEA 4270 CZK
APOSOR901881-5G
3-Acetamido-6-chloropyridazine 98%
3-Acetamido-6-chloropyridazine
Vzorec: C₆H₆ClN₃O
MW: 171,584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835313
CAS číslo: 14959-31-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR