Tisk…

3,5-dichloro-2-nitropyridine 98%

Dodavatel: Apollo Scientific

Synonyms: 3,5-Dichloro-2-nitropyridine

OR901858-1G OR901858-250MG OR901858-5G
APOSOR901858-1GEA 3440 CZK
APOSOR901858-1G APOSOR901858-250MG APOSOR901858-5G
3,5-dichloro-2-nitropyridine 98%
3,5-dichloro-2-nitropyridine
Vzorec: C₅H₂Cl₂N₂O₂
MW: 192,988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254585
CAS číslo: 610278-88-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR