Tisk…

2,6-Dimethyl-3-nitro-4-pyridinol 97%

Dodavatel: Apollo Scientific

Synonyms: 2,6-Dimethyl-3-nitropyridin-4-ol

OR901841-5G
APOSOR901841-5GEA 3410 CZK
APOSOR901841-5G
2,6-Dimethyl-3-nitro-4-pyridinol 97%
2,6-Dimethyl-3-nitro-4-pyridinol
Vzorec: C₇H₈N₂O₃
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18253578
CAS číslo: 13603-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR