Tisk…

Methyl 5-amino-6-chloropyridine-3-carboxylate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-amino-6-chloronicotinate

OR901819-1G OR901819-250MG
APOSOR901819-1GEA 2030 CZK
APOSOR901819-1G APOSOR901819-250MG
Methyl 5-amino-6-chloropyridine-3-carboxylate 98%
Methyl 5-amino-6-chloropyridine-3-carboxylate
Vzorec: C₇H₇ClN₂O₂
MW: 186,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02180841
CAS číslo: 211915-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR