Tisk…

Methyl 6-amino-3-chloropicolinate 95%

Dodavatel: Apollo Scientific

OR901798-1G OR901798-250MG
APOSOR901798-1GEA 2530 CZK
APOSOR901798-1G APOSOR901798-250MG
Methyl 6-amino-3-chloropicolinate 95%
Methyl 6-amino-3-chloropicolinate
Vzorec: C₇H₇ClN₂O₂
MW: 186,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257959
CAS číslo: 1256835-20-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR