3,5-Dichloro-6-methylpyridin-2-ol 98%
Dodavatel: Apollo Scientific
OR901782-1G
OR901782-5G
APOSOR901782-1GEA
4400
CZK
APOSOR901782-1G
APOSOR901782-5G
3,5-Dichloro-6-methylpyridin-2-ol 98%
3,5-Dichloro-6-methylpyridin-2-ol
Vzorec:
C₆H₅Cl₂NO MW: 178.016 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18783194 CAS číslo: 22109-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...