Tisk…

6-Methoxypyridine-2-carbonitrile 95%

Dodavatel: Apollo Scientific

OR901728-5G
APOSOR901728-5GEA 4570 CZK
APOSOR901728-5G
6-Methoxypyridine-2-carbonitrile 95%
6-Methoxypyridine-2-carbonitrile
Vzorec: C₇H₆N₂O
MW: 134,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062856
CAS číslo: 83621-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR