Tisk…
N-tert-Butylnicotinamide 96%
Dodavatel: Apollo Scientific
Synonyms: N-(tert-Butyl)nicotinamide
OR901670-1G OR901670-25G OR901670-5G
APOSOR901670-1GEA 3540 CZK
APOSOR901670-1G APOSOR901670-25G APOSOR901670-5G
N-tert-Butylnicotinamide 96%
N-tert-Butylnicotinamide
Vzorec: C₁₀H₁₄N₂O
MW: 178.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00707839
CAS číslo: 15828-08-7