Tisk…

2-Isopropoxypyridine 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Isopropyloxypyridine

OR901593-1G OR901593-5G
APOSOR901593-1GEA 1450 CZK
APOSOR901593-1G APOSOR901593-5G
2-Isopropoxypyridine 98%
2-Isopropoxypyridine
Vzorec: C₈H₁₁NO
MW: 137.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113404
CAS číslo: 16096-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR