Tisk…

5-Methoxy-1-methyl-benzimidazole-2-carbaldehyde 96%

Dodavatel: Apollo Scientific

OR901582-1G OR901582-250MG
APOSOR901582-1GEA 13660 CZK
APOSOR901582-1G APOSOR901582-250MG
5-Methoxy-1-methyl-benzimidazole-2-carbaldehyde 96%
5-Methoxy-1-methyl-benzimidazole-2-carbaldehyde
Vzorec: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11215482
CAS číslo: 53004-19-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR