(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR900945-1G
OR900945-250MG
APOSOR900945-1GEA
18190
CZK
APOSOR900945-1G
APOSOR900945-250MG
(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester 97%
(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester
Vzorec:
C₁₄H₂₀BNO₃ MW: 261,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18732747 CAS číslo: 631911-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...