Methyl 3-oxo-2-phenylpropanoate 95%
Dodavatel: Apollo Scientific
OR900801-1G
OR900801-5G
APOSOR900801-1GEA
7670
CZK
APOSOR900801-1G
APOSOR900801-5G
Methyl 3-oxo-2-phenylpropanoate 95%
Methyl 3-oxo-2-phenylpropanoate
Vzorec:
C₁₀H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00236385 CAS číslo: 5894-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...