Tisk…

tert-Butyl 3-(methylamino)propionate 97%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 3-(methylamino)propanoate

OR900796-1G OR900796-5G
APOSOR900796-1GEA 5630 CZK
APOSOR900796-1G APOSOR900796-5G
tert-Butyl 3-(methylamino)propionate 97%
tert-Butyl 3-(methylamino)propionate
Vzorec: C₈H₁₇NO₂
MW: 159,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275841
CAS číslo: 143707-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR