Tisk…

[1-(4-Chlorophenyl)cyclopropyl]methanamine 97%

Dodavatel: Apollo Scientific

OR900775-1G OR900775-250MG OR900775-5G
APOSOR900775-1GEA 7050 CZK
APOSOR900775-1G APOSOR900775-250MG APOSOR900775-5G
[1-(4-Chlorophenyl)cyclopropyl]methanamine 97%
[1-(4-Chlorophenyl)cyclopropyl]methanamine
Vzorec: C₁₀H₁₂ClN
MW: 181,662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213095
CAS číslo: 69385-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR