[2-Amino-1-(4-chlorophenyl)ethyl]dimethylamine 97%
Dodavatel: Apollo Scientific
OR900660-1G
OR900660-250MG
APOSOR900660-1GEA
9920
CZK
APOSOR900660-1G
APOSOR900660-250MG
[2-Amino-1-(4-chlorophenyl)ethyl]dimethylamine 97%
[2-Amino-1-(4-chlorophenyl)ethyl]dimethylamine
Vzorec:
C₁₀H₁₅ClN₂ MW: 198,693 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08667746 CAS číslo: 927965-80-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...