Tisk…

[2-Amino-1-(3-chlorophenyl)ethyl]dimethylamine 96%

Dodavatel: Apollo Scientific

OR900656-1G OR900656-250MG
APOSOR900656-1GEA 18420 CZK
APOSOR900656-1G APOSOR900656-250MG
[2-Amino-1-(3-chlorophenyl)ethyl]dimethylamine 96%
[2-Amino-1-(3-chlorophenyl)ethyl]dimethylamine
Vzorec: C₁₀H₁₅ClN₂
MW: 198,692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09262078
CAS číslo: 96798-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR