Tisk…

(1-Methylcyclohexyl)methanamine 96%

Dodavatel: Apollo Scientific

OR900634-1G OR900634-5G
APOSOR900634-1GEA 6220 CZK
APOSOR900634-1G APOSOR900634-5G
(1-Methylcyclohexyl)methanamine 96%
(1-Methylcyclohexyl)methanamine
Vzorec: C₈H₁₇N
MW: 127,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607978
CAS číslo: 3913-98-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR