Tisk…

3-Isopropoxytoluene 90%

Dodavatel: Apollo Scientific

OR900616-10G OR900616-1G OR900616-5G OR900616-25G OR900616-100G
APOSOR900616-10GEA 0 CZK
APOSOR900616-10G APOSOR900616-1G APOSOR900616-5G APOSOR900616-25G APOSOR900616-100G
3-Isopropoxytoluene 90%
3-Isopropoxytoluene
Vzorec: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD20390255
CAS číslo: 19177-04-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR