Tisk…

1-Hydroxycyclopentane-1-carbonitrile 95%

Dodavatel: Apollo Scientific

OR900580-1G OR900580-250MG
APOSOR900580-1GEA 12400 CZK
APOSOR900580-1G APOSOR900580-250MG
1-Hydroxycyclopentane-1-carbonitrile 95%
1-Hydroxycyclopentane-1-carbonitrile
Vzorec: C₆H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00043764
CAS číslo: 5117-85-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR