Tisk…

3-(2-Methylpropoxy)propanenitrile 98%

Dodavatel: Apollo Scientific

OR900475-1G OR900475-5G
APOSOR900475-1GEA 6220 CZK
APOSOR900475-1G APOSOR900475-5G
3-(2-Methylpropoxy)propanenitrile 98%
3-(2-Methylpropoxy)propanenitrile
Vzorec: C₇H₁₃NO
MW: 127,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06252351
CAS číslo: 58936-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR