Tisk…

1-(Methylamino)cyclopentane-1-carbonitrile 95%

Dodavatel: Apollo Scientific

OR900473-1G OR900473-5G
APOSOR900473-1GEA 7670 CZK
APOSOR900473-1G APOSOR900473-5G
1-(Methylamino)cyclopentane-1-carbonitrile 95%
1-(Methylamino)cyclopentane-1-carbonitrile
Vzorec: C₇H₁₂N₂
MW: 124.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040167
CAS číslo: 55793-49-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR