Tisk…

1-Aminocyclopentane-1-carbonitrile 96%

Dodavatel: Apollo Scientific

OR900461-1G OR900461-250MG
APOSOR900461-1GEA 11410 CZK
APOSOR900461-1G APOSOR900461-250MG
1-Aminocyclopentane-1-carbonitrile 96%
1-Aminocyclopentane-1-carbonitrile
Vzorec: C₆H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD05262933
CAS číslo: 49830-37-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR