Tisk…

Methyl 3-formamido-4-hydroxybenzoate 95%

Dodavatel: Apollo Scientific

OR900447-1G OR900447-5G
APOSOR900447-1GEA 8270 CZK
APOSOR900447-1G APOSOR900447-5G
Methyl 3-formamido-4-hydroxybenzoate 95%
Methyl 3-formamido-4-hydroxybenzoate
Vzorec: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD20676665
CAS číslo: 1379261-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR