2-Amino-N-benzylacetamide 96%
Dodavatel: Apollo Scientific
OR900400-1G
OR900400-5G
APOSOR900400-1GEA
11310
CZK
APOSOR900400-1G
APOSOR900400-5G
2-Amino-N-benzylacetamide 96%
2-Amino-N-benzylacetamide
Vzorec:
C₉H₁₂N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD09724204 CAS číslo: 39796-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...