4-(Cyclopropylmethoxy)benzonitrile 98%
Dodavatel: Apollo Scientific
OR900345-1G
OR900345-25G
OR900345-5G
APOSOR900345-1GEA
4140
CZK
APOSOR900345-1G
APOSOR900345-25G
APOSOR900345-5G
4-(Cyclopropylmethoxy)benzonitrile 98%
4-(Cyclopropylmethoxy)benzonitrile
Vzorec:
C₁₁H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD11136694 CAS číslo: 1019619-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...