Tisk…

4-(2-Methylpropoxy)benzonitrile 98%

Dodavatel: Apollo Scientific

Synonyms: 4-iso-Butoxybenzonitrile

OR900341-1G OR900341-5G
APOSOR900341-1GEA 1160 CZK
APOSOR900341-1G APOSOR900341-5G
4-(2-Methylpropoxy)benzonitrile 98%
4-(2-Methylpropoxy)benzonitrile
Vzorec: C₁₁H₁₃NO
MW: 175,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08274895
CAS číslo: 5203-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR