4-(Ethylamino)benzonitrile 98%
Dodavatel: Apollo Scientific
OR900250-1G
OR900250-5G
APOSOR900250-1GEA
1540
CZK
APOSOR900250-1G
APOSOR900250-5G
4-(Ethylamino)benzonitrile 98%
4-(Ethylamino)benzonitrile
Vzorec:
C₉H₁₀N₂ MW: 146,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11135503 CAS číslo: 4714-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...