Tisk…

Chromane 96%

Dodavatel: Apollo Scientific

Synonyms: Chroman

OR900234-1G OR900234-5G
APOSOR900234-1GEA 3060 CZK
APOSOR900234-1G APOSOR900234-5G
Chromane 96%
Chromane
Vzorec: C₉H₁₀O
Storage Temperature: Ambient
MDL Number: MFCD00138123
CAS číslo: 493-08-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR