Tisk…

5-Chloro-7-nitro-1,3-benzoxazole 97%

Dodavatel: Apollo Scientific

OR900191-1G OR900191-250MG
APOSOR900191-1GEA 21490 CZK
APOSOR900191-1G APOSOR900191-250MG
5-Chloro-7-nitro-1,3-benzoxazole 97%
5-Chloro-7-nitro-1,3-benzoxazole
Vzorec: C₇H₃ClN₂O₃
MW: 198,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909485
CAS číslo: 1356111-15-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR