Tisk…
1,4-Dithia-7-azaspiro[4.4]nonane hydrochloride 95%
Dodavatel: Apollo Scientific
OR900152-1G OR900152-250MG
APOSOR900152-1GEA 15110 CZK
APOSOR900152-1G APOSOR900152-250MG
1,4-Dithia-7-azaspiro[4.4]nonane hydrochloride 95%
1,4-Dithia-7-azaspiro[4.4]nonane hydrochloride
Vzorec: C₆H₁₁NS₂·ClH
MW: 197,749 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036094
CAS číslo: 958451-81-5