5-Methyl-2-piperazinobenzonitrile, tech
Dodavatel: Apollo Scientific
|
Varování
|
OR900055-1G
OR900055-5G
APOSOR900055-1GEA
4340
CZK
APOSOR900055-1G
APOSOR900055-5G
5-Methyl-2-piperazinobenzonitrile, tech
5-Methyl-2-piperazinobenzonitrile
Vzorec:
C₁₂H₁₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD18311876 CAS číslo: 1211529-31-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...