tert-Butyl 4-(2-cyano-4-methylphenyl)piperazine-1-carboxylate 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR900048-1G
OR900048-250MG
APOSOR900048-1GEA
13530
CZK
APOSOR900048-1G
APOSOR900048-250MG
tert-Butyl 4-(2-cyano-4-methylphenyl)piperazine-1-carboxylate 96%
tert-Butyl 4-(2-cyano-4-methylphenyl)piperazine-1-carboxylate
Vzorec:
C₁₇H₂₃N₃O₂ MW: 301.383 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17214276 CAS číslo: 1027911-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...